Boats, ceramic, AL23
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
33171.KF 33172.KF 33173.KF 33174.KF 33176.KF 33179.KF 33180.KF 33181.KF
33171.KFEA 60.9 EUR
33171.KF 33172.KF 33173.KF 33174.KF 33176.KF 33179.KF 33180.KF 33181.KF
Boats, ceramic, AL23
Schuitjes Verbrandingsschuitjes
Pure Al₂O₃ Aluminium oxide AL23.
Dense, excellent thermal and electrical resistance properties, corrosion resistant, permeable to microwaves.