Crystal violet lactone 97% (dye content), Sigma-Aldrich®

Leverancier: SIGMA ALDRICH MICROSCOPY
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

332488-25G 332488-5G
332488-25GEA 630 EUR
332488-25G 332488-5G
Crystal violet lactone 97% (dye content), Sigma-Aldrich®
Crystal violet lactone

Crystal Violet lactone, Dye content 97%.


Crystal Violet lactone (CVL) is a colorless lactone derivative of crystal violet. The CVL compound contains two dimethylaniline groups attached via a tetrahedral carbon atom to 6-dimethylaminophthalide at the third position. Crystal violet lactone is suitable for use in color-changing printing. It serves as a reversible color system with temperature change.

Formule: C₂₆H₂₉N₃O₂
Bewaar temperatuur: Room Temperature
MDL Nummer: MFCD00070611
CAS nummer: 1552-42-7
EINECS: 216-293-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=07a9d34a0f97efd23bc5d16a07349e188232ddb4&vid=07a9d34a0f97efd23bc5d16a07349e188232ddb4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance (Form) Powder
Infrared Spectrum Conforms to Structure
Wavelength, c = 0.01 g/L; Methanol 269 - 275 nm
Extinction Coefficient ≥50000
Carbon 72.4 - 77.8%
Nitrogen 9.6 - 10.5%

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR