Zwavelig zuur (≥ 6,0% SO₂) in aqueous solution ACS

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Sulphur dioxide solution

33275.AP 33275.AY
33275.APEA 90.4 EUR
33275.AP 33275.AY
Zwavelig zuur (≥ 6,0% SO₂) in aqueous solution ACS
Zwavelig zuur

CAS No.: 7782-99-2 Molecular Formula: H2SO3 Formula Weight: 82.08 Physical Form: Liquid Density: 1.030 Sensitivity: Air Sensitive Appearance: Colourless Odour: Pungent Merck Reference: 14,8976 UN No.: 1833 MDL No.: MFCD00011335

Formule: H₂O₃S
MW: 82.08 g/mol
Dichtheid: 1.030
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011335
CAS nummer: 7782-99-2
EINECS: 231-973-1
UN: 1833
ADR: 8 II
Merck Index: 14,8976

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR