Cyclohexanon 99+% ACS
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
Waarschuwing
33309.AP 33309.K2 33309.K7 33309.M1
33309.APEA 45.3 EUR
33309.AP 33309.K2 33309.K7 33309.M1
Cyclohexanon 99+% ACS
Cyclohexanon
Purity: 99+% CAS No.: 108-94-1 Molecular Formula: C6H10O Formula Weight: 98.15 Flash Point: 47°(116°F) Physical Form: Liquid Density: 0.9478 Refractive Index: 1.4507 Melting Point: -26° Boiling Point: 154-156° Beilstein Reference: 385735 Merck Reference: 14,2726 UN No.: 1915 MDL No.: MFCD00001625
Formule: C₆H₁₀(=O)
Kookpunt: 154…156 °C
Smeltpunt: –26 °C
Dichtheid: 0,9478
Vlampunt: 47 °C (116 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00001625
CAS nummer: 108-94-1
EINECS: 203-631-1
UN: 1915
ADR: 3,III
Bereik: 01-2119453616-35
Merck Index: 14,2726
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b8701dac7edb70a452483cd4f1bd716ca29ba9aa&vid=0360daffe7606174162954ba58a6ba2b78e28dfb&prid=web&clid=SAPDAM