Calciumchloride, watervrij ≥93%, porous

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

33327.36 33327.A3
33327.36EA 47.7 EUR
33327.36 33327.A3
Calciumchloride, watervrij ≥93%, porous
Calciumchloride

Purity: 93% CAS No.: 10043-52-4 Molecular Formula: CaCl2 Formula Weight: 110.99 Physical Form: -4 Mesh Sensitivity: Hygroscopic Boiling Point: 1600° Merck Reference: 14,1659 MDL No.: MFCD00010903

Formule: CaCl₂
Kookpunt: >1600 °C
Smeltpunt: 782 °C
Dichtheid: 2,174 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00010903
CAS nummer: 10043-52-4
EINECS: 233-140-8
Bereik: 01-2119494219-28
Merck Index: 14,1659

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR