Manganese(II) sulphate monohydrate 98.0-101.0% ACS

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

33341.36 33341.A3
33341.36EA 51.2 EUR
33341.36 33341.A3
Manganese(II) sulphate monohydrate 98.0-101.0% ACS
Manganese(II) sulphate monohydrate

Purity: 98.0-101.0% CAS No.: 10034-96-5 Molecular Formula: MnSO4.H2O Formula Weight: 169.02 (151.00anhy) Physical Form: Powder Density: 2.95 pH: 3-3.5 Merck Reference: 14,5739 UN No.: 3077 MDL No.: MFCD00149159

Formule: MnSO₄·1H₂O
Smeltpunt: 700 °C
Dichtheid: 2,95
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149159
CAS nummer: 10034-96-5
EINECS: 232-089-9
UN: 3077
ADR: 9,III
Merck Index: 14,5739

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR