4-Methyl-2-pentaanon ≥98.5% ACS

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Methyl iso-Butyl Ketone, Isobutyl Methyl Ketone, Methyl Isobutyl Ketone, MIK

33346.AP 33346.K2
33346.APEA 41.2 EUR
33346.AP 33346.K2
4-Methyl-2-pentaanon ≥98.5% ACS
4-Methyl-2-pentaanon

Purity: 98.5+% CAS No.: 108-10-1 Molecular Formula: C6H12O Formula Weight: 100.16 Flash Point: 13°(55°F) Physical Form: Liquid Density: 0.801 Refractive Index: 1.3960 Appearance: Colourless Odour: Mild Melting Point: -84° Boiling Point: 117-118° Beilstein Reference: 605399 Merck Reference: 14,5207 UN No.: 1245 MDL No.: MFCD00008938

Formule: (CH₃)₂CHCH₂COCH₃
MW: 100.16 g/mol
Kookpunt: 117…118 °C
Smeltpunt: –84 °C
Dichtheid: 0.801
Vlampunt: 13 °C (55 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008938
CAS nummer: 108-10-1
EINECS: 203-550-1
UN: 1245
ADR: 3 II
Merck Index: 14,5207
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3d44947ef43cbc8103ae523ce5da380ef184ba09&vid=373b9075524004b95d32d3cf9698896714467e8c&prid=web&clid=SAPDAM

Order Now

Specificaties

Environmentally Preferable Veiligere gezondheid van mens en milieu
Milieu certificering Chem21 Solvent Selection Guide recommended by default and after discussion

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR