N-(1-Naphthyl)ethylenediamine dihydrochloride, Supelco®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Marshall's Reagent, N-2-Aminoethyl-1-naphthylamine dihydrochloride, N-1-Naphthylethylenediamine

M-Clarity™ quality level = MQ200

33461-25G 33461-5G
33461-25GEA 235 EUR
33461-25G 33461-5G
N-(1-Naphthyl)ethylenediamine dihydrochloride, Supelco®
N-(1-Naphthyl)ethylenediamine dihydrochloride
Formule: C₁₂H₁₆Cl₂N₂
MW: 259.18 g/mol
Smeltpunt: 188…190 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00012556
CAS nummer: 1465-25-4
EINECS: 215-981-2
Merck Index: 12,06496
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a5286d359f2420bda1d50e99536d4fd026e2aafd&vid=b80fc9486585c357d437c2db62e38327db5fc294&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR