Food standards, T-2 Toxin, neat, Supelco®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
33947-5MG
33947-5MGEA 487 EUR
33947-5MG
Food standards, T-2 Toxin, neat, Supelco®
Standaarden Standaarden voor de voedselindustrie
T-2 Toxin is found to be a potent trichothecene mycotoxin produced by the genus fusarium fungi in feedstuffs and cereal grains.
Opgelet: Storage temperature: Freezer −20 °C
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3272c10a2ae13c22296f0b151cdc25420b989bd4&vid=3272c10a2ae13c22296f0b151cdc25420b989bd4&prid=web&clid=SAPDAM