Organic reference standard, Nivalenol solution, analytical standard, Supelco®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
34131-2ML
34131-2MLEA 453 EUR
34131-2ML
Organic reference standard, Nivalenol solution, analytical standard, Supelco®
Standaarden Organische standaarden
Nivalenol is a potential trichothecene mycotoxin, produced by Fusarium nivale.
Genotoxicity of nivalenol is evaluated by alkaline single-cell gel electrophoresis assay.
Nivalenol is a secondary metabolite produced by Fusarium species, which is a plant pathogenic fungus of wheat and other cereals.
Nivalenol was used as standard to investigate in vitro mycotoxin binding of deoxynivalenol and nivalenol by adsorbent materials (activated carbon).
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d51b213e2468c210b10e1f913fab6bac2d071607&vid=d51b213e2468c210b10e1f913fab6bac2d071607&prid=web&clid=SAPDAM