Calbiochem® Bovine native Fibrinogen (from Plasma)
Leverancier: Merck Millipore Calbiochem
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
341573-1
341573-1EA 310 EUR
341573-1
Calbiochem® Bovine native Fibrinogen (from Plasma)
Eiwitten en peptiden
Fibrinogen is a soluble plasma glycoprotein that is synthesised in the hepatic parenchymal cells.
  • Contains more than 95% clottable protein