Eosin Y (yellowish), Gurr™ for microscopical staining

Leverancier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Solvent Red 43, Field's Stain B, Eosin 225, 2,4,5,7-Tetrabromofluorescein disodium salt, Disodium 2-(2,4,5,7-tetrabromo-6-oxido-3-oxoxanthen-9-yl)benzoate, Disodium 2',4',5',7'-tetrabromo-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-3',6'-bis(olate), Tetrabromofluorescein disodium salt, 2′,4′,5′,7′-Tetrabromofluorescein disodium salt, Eosin Y disodium salt

341975TEA 1570 EUR
341975T
Eosin Y (yellowish), Gurr™ for microscopical staining
Eosin Y (yellowish)
Formule: C₂₀H₆Br₄Na₂O₅
MDL Nummer: MFCD00005040
CAS nummer: 17372-87-1
IVDLogo

Order Now

Specification Test Results

TLC-Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR