Zwavelkoolstof 99,9, reagent Benzene_low

Leverancier: Honeywell Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

342270-100ML 342270-1L 342270-800ML
342270-100MLEA 53.7 EUR
342270-100ML 342270-1L 342270-800ML
Zwavelkoolstof 99,9, reagent Benzene_low
Zwavelkoolstof

  • ≥95% for general laboratory use.

Formule: CS₂
MW: 76.14 g/mol
Kookpunt: 46 °C (1.013 hPa)
Smeltpunt: –76 °C
Dichtheid: 1.264 g/cm³ (20 °C)
Vlampunt: –30 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011321
CAS nummer: 75-15-0
EINECS: 200-843-6
UN: 1131
ADR: 3 I
Merck Index: 13,01821
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a7fe0c906449b3b5461b1bac971c583c92c0b41c&vid=59e9028fbb4d1c2acb6f98101726e8ec520e0aa8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Explosion Limit 50 %
Assay (GC) Min. 99.90 %
Water (Karl Fischer) Max. 0.05 %
Non-volatile matter Max. 0.0003 %
Benzene (GC) Max. 1 ppm
Autoignition Temperature 212 °F
Vapor Density 2.67 (vs air)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR