CyMet KX Haematology CN free

Leverancier: Avantor
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

3425.0500EA 43.57 EUR
3425.0500
CyMet KX Haematology CN free
CyMet

Lysing reagent for hematology


Opgelet: In vitro diagnostic medical devices only for professional use.

IVDLogo

Order Now

Specification Test Results

Conductivity (mS) 8.2 - 9.2
Osmolality (mOsm/kg) 129 - 139
Performance Test passes test
pH at 25°C 4.4 - 4.6
*) All parameters except mid cells: Test/Ref ratio > 95% and < 105%. Correlation coefficient: > 0.95


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR