Methyl red 0,2 g/l ethanol, TECHNICAL

Leverancier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: 2-(4-Dimethylaminophenylazo)benzoic acid, 2-((4-(Dimethylamino)phenyl)diazenyl)benzoic acid

34302.231EA 60.6 EUR
34302.231
Methyl red 0,2 g/l ethanol, TECHNICAL
Methyl red

  • Transition range: pH 4,4 - 6,2

Formule: C₁₅H₁₅N₃O₂
MW: 269.3 g/mol
Vlampunt: 20 °C (gesloten beker)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002425
CAS nummer: 493-52-7
EINECS: 207-776-1
UN: 1170
ADR: 3 II
Merck Index: 12,06199

Order Now

Specification Test Results

Identification Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR