Ferroin (1,10-phenanthrolineferrous sulphate complex) solution Reag. Ph. Eur. redox indicator E0 (H2SO4 1 mol/l) = +1,06 volt, Fluka™

Leverancier: Honeywell Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Tris(1,10-phenanthroline)iron (II) sulphate, Tris(1,10-phenanthroline-N1,N10)iron sulphate

34531-100ML 34531-500ML
34531-100MLEA 65.6 EUR
34531-100ML 34531-500ML
Ferroin (1,10-phenanthrolineferrous sulphate complex) solution Reag. Ph. Eur. redox indicator E0 (H2SO4 1 mol/l) = +1,06 volt, Fluka™
Ferroin (1,10-phenanthrolineferrous sulphate complex)

E0 in sulfuric acid 1 mol/l = +1,06 volt

Formule: C₃₆H₂₄FeN₆SO₄
Kookpunt: 100 °C (1,013 hPa)
Dichtheid: 1,00 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00036428
CAS nummer: 14634-91-4

Order Now

Specification Test Results

Change complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR