Genistein, Millipore®

Leverancier: Merck Millipore Calbiochem
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

345834-20 345834-50
345834-20EA 143 EUR
345834-20 345834-50
Genistein, Millipore®
Genistein
Formule: C₁₅H₁₀O₅
Smeltpunt: 297…298 °C
MDL Nummer: MFCD00016952
CAS nummer: 446-72-0
EINECS: 207-174-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR