Mierenzuur 100%, CHROMASOLV™ LC-MS LabReady™, Solvent Blend, Riedel-de Haën™

Leverancier: Honeywell Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

34668-2.5L 34668-4X4L
34668-2.5LEA 284 EUR
34668-2.5L 34668-4X4L
Mierenzuur 100%, CHROMASOLV™ LC-MS LabReady™, Solvent Blend, Riedel-de Haën™
Mierenzuur

  • For LC-MS and LC-UV trace metals analysis
  • HPLC / UV-VIS quality control
  • Low gradient baseline drift
  • Low levels of trace metals
  • No non-volatile impurities

Formule: HCO₂H
Kookpunt: 82 °C (1,013 hPa)
Smeltpunt: –46 °C
Dichtheid: 0,7767 g/cm³ (25 °C)
Vlampunt: 8 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00001878
CAS nummer: 64-18-6
EINECS: 200-835-2
UN: 1648
ADR: 3,II

Order Now

Specification Test Results

Assay (GC) Min. 99.5 %
Assay of formic acid 0.093 - 0.107 %
Transmittance at 254 nm Min. 90 %
Transmittance at 230 nm Min. 15 %
Transmittance at 210 nm Min. 5 %
Suitability for the LC-MS complying
HPLC Gradient at 254 nm Max. 50 mAU
Calcium (Ca) Max. 0.5 ppm
Potassium (K) Max. 0.5 ppm
Magnesium (Mg) Max. 0.5 ppm
Sodium (Na) Max. 2 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR