Aceton ≥99.8%, CHROMASOLV™ for HPLC, Riedel-de Haën™

Leverancier: Honeywell Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

34850-45L 34850-1L 34850-2.5L 34850-4X2.5L 34850-6X1L 34850-18L 34850-190LRC 34850-190L-RC
34850-45LEA 3080 EUR
34850-45L 34850-1L 34850-2.5L 34850-4X2.5L 34850-6X1L HONC34850-18L HONC34850-190LRC HONC34850-190L-RC
Aceton ≥99.8%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Aceton

  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency

Formule: CH₃COCH₃
Kookpunt: 55…57 °C (1,013 hPa)
Smeltpunt: –95 °C
Dichtheid: 0,79 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008765
CAS nummer: 67-64-1
EINECS: 200-662-2
UN: 1090
ADR: 3,II
Bereik: 01-2119471330-49
Merck Index: 13,00067
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3de5d8e3bf7a94692d684c8f0ccdfb8500fde76d&vid=24fb2e0ec86974b1803780c7d5b23804a085df64&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) Min. 99.8 %
Non-volatile matter Max. 0.0005 %
Water (Karl Fischer) Max. 0.1 %
Free acid (as CH₃COOH) Max. 0.002 %
Transmittance at 330 nm Min. 15 %
Transmittance at 335 nm Min. 50 %
Transmittance at 340 nm Min. 80 %
Transmittance from 350 nm Min. 98 %
Vapor Density 2 (vs air)
Vapor Pressure 184 mmHg ( 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR