1-Propanol ≥99.9%, CHROMASOLV™ for HPLC, Riedel-de Haën™

Leverancier: Honeywell Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Propylic alcohol, Propan-1-ol, n-Propan-1-ol, Ethylcarbinol, n-Propyl alcohol, Propanol-1, n-Propanol, Propyl alcohol, 1-Propyl alcohol, 1-Hydroxypropane

34871-1L 34871-2.5L 34871-4X2.5L 34871-6X1L
34871-1LEA 155 EUR
34871-1L 34871-2.5L 34871-4X2.5L 34871-6X1L
1-Propanol ≥99.9%, CHROMASOLV™ for HPLC, Riedel-de Haën™
1-Propanol

  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency


Beilstein Number: 1098242

Formule: H₃CCH₂CH₂OH
Kookpunt: 96-98 °C (1,013 hPa)
Smeltpunt: -127 °C
Dichtheid: 0,803…0.804 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002941
CAS nummer: 71-23-8
EINECS: 200-746-9
UN: 1274
ADR: 3,II
Merck Index: 13,07934
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ab459f809f7797b46eb6d275aa3b1e6e94dc70d1&vid=deb28b985024604344065eea93ff6c6a9dd5a431&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) Min. 99.9 %
Non-volatile matter Max. 0.0005 %
Water (Karl Fischer) Max. 0.05 %
Free acid (as C₂H₅COOH) Max. 0.001 %
Transmittance at 220 nm Min. 40 %
Transmittance at 240 nm Min. 85 %
Transmittance from 275 nm Min. 99 %
Vapor Pressure 10 mmHg ( 147 °C)
Autoignition Temperature 700 °F
Vapor Density 2.1 (vs air)
Refractive Index n20/D 1.384 (lit.)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR