Anti-CD8A Rat Monoclonal Antibody (FITC (Fluorescein)) [clone: 53-6.7]
Leverancier: Tonbo Biosciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
35-0081-U500 35-0081-U100 35-0081-U025
35-0081-U500EA 104 EUR
35-0081-U500 35-0081-U100 35-0081-U025
Anti-CD8A Rat Monoclonal Antibody (FITC (Fluorescein)) [clone: 53-6.7]
Antilichamen
The 53-6.7 antibody reacts with the 32-34 kDa alpha subunit of mouse CD8, known as CD8a or CD8 alpha. CD8a can form a homodimer (CD8 alpha-alpha), but is more commonly expressed as a heterodimer with a second chain known as CD8b or CD8 beta. CD8 acts as a co-receptor in antigen recognition and subsequent T cell activation that is initiated upon binding of the T cell receptor (TCR) to antigen-bearing MHC Class I molecules. The cytoplasmic domains of CD8 provide binding sites for the tyrosine kinase lck, facilitating intracellular signaling events that lead to T cell activation, development,More Product Information