Anti-CD20 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 2H7]

Leverancier: Tonbo Biosciences
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

35-0209-T025 35-0209-T100 35-0209-T500
35-0209-T025EA 51.2 EUR
35-0209-T025 35-0209-T100 35-0209-T500
Anti-CD20 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 2H7]
Antilichamen
The 2H7 antibody reacts with human CD20, expressed by pre-B cells, resting and activated B cells, follicular dendritic cells and a small subset of T cells, but not on plasma cells. CD20 is a 33-37 kDa transmembrane protein which is differentially phosphorylated in resting and active B cells. The CD20 molecule plays a role in regulating B-cell activation.

Type: Primary
Antigen: CD20
Clonality: Monoclonal
Clone: 2H7
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
Order Now

Specificaties

antigeen symbool MS4A1
Environmentally Preferable
Type antilichaam Primair,Membrane-spanning 4-Domains, Subfamily A, Member 1
Antigeen synoniemen CVID5|Bp35|S7|LEU-16|MS4A2|CD20|B1
Type antilichaam Primair,Membrane-spanning 4-Domains, Subfamily A, Member 1
Clonality Monoklonaal
Conjugatie FITC (Fluoresceïne-isothiocyanaat)
Clone 2H7
Reactivity Human
Host Mouse
Gen-ID 931
Isotype IgG2b kappa
Flowcytometrie Yes
Kruisadsorptie No
Opslagtemperatuur Store undiluted at 2-8 °C, protect from light, do not freeze.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR