Human processed serum (HPS), T3/T4 depleted

Leverancier: SEQENS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

35000D-100ML 35000D-500ML 35000D-1L
35000D-100MLEA 255 EUR
35000D-100ML 35000D-500ML 35000D-1L
Human processed serum (HPS), T3/T4 depleted
Serum
SEQENS IVD manufactures T3/T4 Depleted Human Processed Serum.

  • Dry extract: 65 to 85 g/L
  • Total proteins: 55 to 75 g/L
  • Total T3: <0,6 nmol/L
  • Total T4: <13 nmol/L
  • Free T3: <1,5 pmol/L
  • Free T4: <5,2 pmol/L
  • Filtered through a 0,2 µm membrane
  • Shelf life: 2 years

For this product, source material has been tested and found negative or non-reactive (FDA approved and/or CE marked test methods) for anti-HIV 1+2 and anti-HCV antibodies, HBs antigen, HIV 1 and HCV RNA and Syphilitic serology.

Complete donor traceability on file.

Opgelet: Only forMore Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR