Natriumthiosufaat 0.1 M Na2S2O3 (0.1N) aqueous solution Reag. Ph. Eur. Volumetric, Fluka™

Leverancier: Honeywell Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Sodiumthiosulphate, Sodium hyposulphite, Antichlor, di-Sodium thiosulphate

35245-1L 35245-10L-VP 35245-5L-VP 35245-6X1L
35245-1LEA 41.7 EUR
35245-1L 35245-10L-VP 35245-5L-VP 35245-6X1L
Natriumthiosufaat 0.1 M Na2S2O3 (0.1N) aqueous solution Reag. Ph. Eur. Volumetric, Fluka™
Natriumthiosufaat
Formule: Na₂S₂O₃
MW: 158.11 g/mol
Kookpunt: 100 °C (1.013 hPa)
Smeltpunt: 48
Dichtheid: 1.0 g/cm³
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003499
CAS nummer: 7772-98-7
EINECS: 231-867-5
Merck Index: 12,08844

Order Now

Specification Test Results

Factor 0.999 - 1.001


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR