Magnesium oxide ≥95.0% ACS

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

35442.22 35442.36 35442.A3
35442.22EA 91.8 EUR
35442.22 35442.36 35442.A3
Magnesium oxide ≥95.0% ACS
Magnesium oxide

Purity: 95.0% min CAS No.: 1309-48-4 Molecular Formula: MgO Formula Weight: 40.30 Physical Form: Granular Density: 3.58 Refractive Index: 1.736 Odour: Odourless Melting Point: 2852° Boiling Point: 3600° Merck Reference: 14,5677 MDL No.: MFCD00011109

Formule: MgO
Kookpunt: 3600 °C
Smeltpunt: 2852 °C
Dichtheid: 3,58
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011109
CAS nummer: 1309-48-4
EINECS: 215-171-9
Merck Index: 14,5677

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR