Calcium fluoride, ultra dry ≥99.99% (metals basis)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing


Ampuled under argon

35742.06 35742.14 35742.22
35742.06EA 128 EUR
35742.06 35742.14 35742.22
Calcium fluoride, ultra dry ≥99.99% (metals basis)
Calcium fluoride

Purity: 99.99% CAS No.: 7789-75-5 Molecular Formula: CaF2 Formula Weight: 78.08 Physical Form: Powder, Ampouled under argon Density: 3.18 Refractive Index: 1.434 Sensitivity: Hygroscopic Odour: Odourless Melting Point: 1403° Boiling Point: ca 2500° Merck Reference: 14,1667 MDL No.: MFCD00010907

Formule: CaF₂
MW: 78.07 g/mol
Kookpunt: ∼2500 °C
Smeltpunt: 1403 °C
Dichtheid: 3.18
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00010907
CAS nummer: 7789-75-5
EINECS: 232-188-7
Merck Index: 14,1667

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR