Scandium standard solution, 10000 mg/L Sc in 5% nitric acid (from Sc₂O₃), standard for ICP, Specpure®

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

35755.AD 35755.AE 35755.AP
35755.ADEA 265 EUR
35755.AD 35755.AE 35755.AP
Scandium standard solution, 10000 mg/L Sc in 5% nitric acid (from Sc₂O₃), standard for ICP, Specpure®
Standaarden Kationische standaarden Scandium standaarden

Molecular Formula: Sc₂O₃ in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00016323


Certificaten: Traceable to NIST

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR