Europium standard solution, 10000 mg/L Eu in 5% nitric acid (from Eu?O?), standard for ICP, Specpure®

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

35761.AD 35761.AE 35761.AP
35761.ADEA 97.1 EUR
35761.AD 35761.AE 35761.AP
Europium standard solution, 10000 mg/L Eu in 5% nitric acid (from Eu?O?), standard for ICP, Specpure®
Standaarden Kationische standaarden Europium standaarden

Molecular Formula: Eu₂O₃ in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00010992


Certificaten: Traceable to NIST

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR