Titanium(IV) isopropoxide ≥99.995% (metals basis)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

35794.06 35794.14 35794.22
35794.06EA 47 EUR
35794.06 35794.14 35794.22
Titanium(IV) isopropoxide ≥99.995% (metals basis)
Titanium(IV) isopropoxide

Purity: 99.995% CAS No.: 546-68-9 Molecular Formula: C12H28O4Ti Formula Weight: 284.23 Flash Point: 46°(115°F) Physical Form: Liquid Density: 0.955 Refractive Index: 1.4640 Sensitivity: Moisture Sensitive Appearance: Colourless Melting Point: 16-20° Boiling Point: 232° Beilstein Reference: 3679474 Merck Reference: 14,9480 UN No.: 2413 MDL No.: MFCD00008871

Formule: C₁₂H₂₈O₄Ti
Kookpunt: 232 °C
Smeltpunt: 16…20 °C
Dichtheid: 0,955
Vlampunt: 46 °C (115 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008871
CAS nummer: 546-68-9
EINECS: 208-909-6
UN: 2413
ADR: 3,III
Merck Index: 14,9480

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR