Lanthanum(III) bromide, ultra dry ≥99.9% (REO, rare earth oxide basis)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing


Ampuled under argon

35800.04 35800.09 35800.18
35800.04EA 117 EUR
35800.04 35800.09 35800.18
Lanthanum(III) bromide, ultra dry ≥99.9% (REO, rare earth oxide basis)
Lanthanum(III) bromide

Purity: 99.9% CAS No.: 13536-79-3 Molecular Formula: LaBr3 Formula Weight: 378.62 Physical Form: -10 Mesh Beads, Ampouled under argon Sensitivity: Hygroscopic MDL No.: MFCD00016161

Formule: Br₃La
Smeltpunt: 789 °C
Dichtheid: 5,06 g/cm³ (25 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00016161
CAS nummer: 13536-79-3
EINECS: 236-896-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR