Copper(II) isopropoxide ≥98% (metals basis)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

35821.06 35821.14 35821.03
35821.06EA 241 EUR
35821.06 35821.14 35821.03
Copper(II) isopropoxide ≥98% (metals basis)
Copper(II) isopropoxide

Purity: 98% CAS No.: 23578-23-6 Molecular Formula: [(CH3)2CHO]2Cu Formula Weight: 181.73 Physical Form: Powder Sensitivity: Moisture Sensitive UN No.: 3180 MDL No.: MFCD00074982

Formule: C₆H₁₄CuO₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00074982
CAS nummer: 23578-23-6
UN: 3181
ADR: 4.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR