Fenol, Supelco®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Phenylalcohol, Carbolic acid, Hydroxybenzene

MQ100

35952-1G
35952-1GEA 82.7 EUR
35952-1G
Fenol, Supelco®
Fenol
Formule: C₆H₅OH
MW: 94.11 g/mol
Kookpunt: 181.7
Smeltpunt: 40.8
Dichtheid: 1.0722 g/cm³ (20 °C)
Vlampunt: 81
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002143
CAS nummer: 108-95-2
EINECS: 203-632-7
UN: 1671
ADR: 6.1 II
Bereik: 01-2119471329-32
Merck Index: 12,07390
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=80f31910b809334768ad738f4e219ef9987ef243&vid=2b5a8fa6b3ea7d7a468d90abf4102ae68393f63f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR