Karl fischer reagents, Fluka™
Leverancier: Honeywell Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
36115-1L 36116-1L 36116-6X1L 36117-1L
36115-1LEA 140 EUR
36115-1L 36116-1L 36116-6X1L 36117-1L
Karl fischer reagents, Fluka™
Karl Fischer titranten en reagentia
Karl fischer reagents can be used for for pyridine-based volumetric KF titration.
  • Titre: 5,5 to 6,5 mg/ml
Mixture of solutions A and B (ratio 1:1) yield water equivalent (titre) of mi, for pyridine-based volumetric KF titration.