Gold ≥99.95% (metals basis), foil, Thickness: 0.5 mm (0.02 in)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

36148.FF 36148.FI
36148.FFEA 1615 EUR
36148.FF 36148.FI
Gold ≥99.95% (metals basis), foil, Thickness: 0.5 mm (0.02 in)
Gold

Purity: 99.95% CAS No.: 7440-57-5 Physical Form: ≈6g/25x25mm Odour: Odourless MDL No.: MFCD00003436

Formule: Au
MW: 196.97 g/mol
Kookpunt: 2800
Smeltpunt: 1063
Dichtheid: 19.3 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003436
CAS nummer: 7440-57-5
EINECS: 231-165-9
Merck Index: 12,04526

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR