Nickel chromium alloy (80:20 w%) ≥99.9% (metals basis), lumps

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

36298.14 36298.22
36298.14EA 115 EUR
36298.14 36298.22
Nickel chromium alloy (80:20 w%) ≥99.9% (metals basis), lumps
Nickel chromium alloy (80:20 w%)

Purity: 99.9% CAS No.: 11106-97-1 Molecular Formula: Ni:Cr; 80:20 wt% Physical Form: 3-12mm Sintered Lump Odour: Odourless


For vacuum deposition

Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00673110
CAS nummer: 11106-97-1

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR