Dimethylsulfoxide ≥99.9% ACS

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Methanesulphinylmethane, Methyl sulphoxide, Dimethyl(oxido)sulphur, DMSO

36480.AP 36480.K2 36480.M1
36480.APEA 56.6 EUR
36480.AP 36480.K2 36480.M1
Dimethylsulfoxide ≥99.9% ACS
Dimethylsulfoxide

Purity: 99.9% min CAS No.: 67-68-5 Molecular Formula: C2H6OS Formula Weight: 78.13 Flash Point: 87°(189°F) Physical Form: Clear liquid Density: 1.101 Refractive Index: 1.4790 Sensitivity: Hygroscopic Appearance: Colourless Odour: Mild Melting Point: 18.4° Boiling Point: 189° Beilstein Reference: 506008 Merck Reference: 14,3259 MDL No.: MFCD00002089

Formule: (CH₃)₂SO
MW: 78.14 g/mol
Kookpunt: 189 °C
Smeltpunt: 18.4 °C
Dichtheid: 1.101
Vlampunt: 87 °C (189 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002089
CAS nummer: 67-68-5
EINECS: 200-664-3
Merck Index: 14,3259
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ec1573e80c11ddf9254283ef76abf5fb6539c7e&vid=157e2a24c93041cbbb69833d226195e333b07665&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR