Tin(IV) isopropoxide 10% (w/v) in 2-propanol/toluene , 98% (metals basis)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

36563.AA 36563.AE
36563.AAEA 41.2 EUR
36563.AA 36563.AE
Tin(IV) isopropoxide 10% (w/v) in 2-propanol/toluene , 98% (metals basis)
Tin(IV) isopropoxide

Purity: 98% CAS No.: 1184-61-8 Molecular Formula: Sn[OCH(CH3)2]4 Formula Weight: 355.06 Physical Form: Liquid Sensitivity: Moisture Sensitive UN No.: UN1993 MDL No.: MFCD00145408

Formule: C₁₂H₂₈O₄Sn
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00145408
CAS nummer: 1184-61-8
UN: 1993
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR