Starch, soluble ACS for iodometric titrations

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

36703.18 36703.30 36703.A1
36703.18EA 37.2 EUR
36703.18 36703.30 36703.A1
Starch, soluble ACS for iodometric titrations
Starch, soluble

CAS No.: 9005-84-9 Physical Form: Powder Odour: Odourless Melting Point: 256-258° dec. Solubility: Soluble in hot water Merck Reference: 14,8799 MDL No.: MFCD00082026

Formule: (C₆H₁₀O₅)n
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00082026
CAS nummer: 9005-84-9
EINECS: 232-686-4
Merck Index: 14,8799

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR