Sodium metaborate tetrahydrate, tech.

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

36736.A1 36736.A7
36736.A1EA 60.6 EUR
36736.A1 36736.A7
Sodium metaborate tetrahydrate, tech.
Sodium metaborate tetrahydrate

CAS No.: 10555-76-7 Molecular Formula: NaBO2.4H2O Formula Weight: 137.86 (65.80anhy) Physical Form: -8 Mesh Crystalline Melting Point: ca 53° dec. MDL No.: MFCD00149205

Formule: BNaO₂·4H₂O
MW: 137.86 g/mol
Smeltpunt: ∼ 53 °C (ontbinding)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149205
CAS nummer: 10555-76-7
EINECS: 231-891-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR