N,N-Dimethylaceetamide, extra dry over Molecular Sieve 99.5%, AcroSeal®

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

375231000 375230010
375231000.EA 34.5 EUR
375231000. 375230010.
N,N-Dimethylaceetamide, extra dry over Molecular Sieve 99.5%, AcroSeal®
N,N-Dimethylaceetamide
Formule: CH₃CON(CH₃)₂
Kookpunt: 164…166 °C (760 mmHg)
Smeltpunt: –20 °C
Dichtheid: 0,937 g/cm³
Vlampunt: 70 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008686
CAS nummer: 127-19-5
EINECS: 204-826-4
ADR: 6.1,III
Merck Index: 13,03254
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ace04c31b8db2d1fc0c18051cd2eefc52a61ea65&vid=4e98bd4c34a9326a56ee3e11797b2e6e1b323c8f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Water ≤0.005 %
GC ≥99.4 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR