Dextran, M.W. ~ 150,000, powder

Leverancier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

381082MEA 225 EUR
381082M 381082M1
Dextran, M.W. ~ 150,000, powder
Dextran
Formule: (C₆H₁₀O₅)n
MDL Nummer: MFCD00130935
CAS nummer: 9004-54-0
EINECS: 232-677-5

Order Now

Specification Test Results

Appearance White/almost white powder
Intrinsic viscosity (37°C) 0.32 - 0.37
Loss on drying Max. 10.0 %
pH range 4.0 - 7.0
Solubility in water (1 %) Passes test
Ashes Max. 2.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR