Zoutzuur concentrate for dilution to 1L, 0.5 M HCl (0.5N) volumetrische oplossing, Fluka™

Leverancier: Honeywell Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

38285-1EA 38285-6X1EA
38285-1EAEA 41.5 EUR
38285-1EA 38285-6X1EA
Zoutzuur concentrate for dilution to 1L, 0.5 M HCl (0.5N) volumetrische oplossing, Fluka™
Zoutzuur

Concentrate for 1L standard solution

Formule: HCl
Dichtheid: 1,11 g/cm³
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011324
CAS nummer: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,II

Order Now

Specification Test Results

Factor 0.998 - 1.002
BAM/EMPA reference material effective
BAM/EMPA Certificate Number effective
Traceable to NIST SRM effective


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR