Zwavelzuur concentrate for dilution to 1L, 0.005 M H2SO4 (0.01N) Volumetric, Fluka™

Leverancier: Honeywell Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

38308-1EA
38308-1EAEA 84.7 EUR
38308-1EA
Zwavelzuur concentrate for dilution to 1L, 0.005 M H2SO4 (0.01N) Volumetric, Fluka™
Zwavelzuur

Concentrate for 1L standard solution

Formule: H₂SO₄
MW: 98.08 g/mol
Kookpunt: 100 °C (1.013 hPa)
Smeltpunt: 10.38
Dichtheid: 1.010 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00064589
CAS nummer: 7664-93-9
EINECS: 231-639-5
UN: 3264
ADR: 8 III

Order Now

Specification Test Results

Factor 0.998 - 1.002
BAM/EMPA reference material effective
BAM/EMPA Certificate Number effective
Traceable to NIST SRM effective


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR