Platinum 0,5% on carbon, granules, reduced

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

38344.06 38344.14
38344.06EA 146 EUR
38344.06 38344.14
Platinum 0,5% on carbon, granules, reduced
Platinum

CAS No.: 7440-06-4 Physical Form: 2-5mm granules Odour: Odourless UN No.: UN1325 MDL No.: MFCD00011179

Formule: Pt
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011179
CAS nummer: 7440-06-4
UN: 1325
ADR: 4.1,III

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR