Strontium ≥99.95% (metals basis), pieces, distilled dendritic

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar


Ampuled under argon

38624.06 38624.14
38624.06EA 841 EUR
38624.06 38624.14
Strontium ≥99.95% (metals basis), pieces, distilled dendritic
Strontium

Purity: 99.95% CAS No.: 7440-24-6 Physical Form: Ampouled under argon Sensitivity: Air and Moisture Sensitive UN No.: 3208 MDL No.: MFCD00134060

Formule: Sr
MW: 87.62 g/mol
Kookpunt: 1151
Smeltpunt: 570…660 °C
Dichtheid: 2.38 g/cm³ (770 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00134060
CAS nummer: 7440-24-6
EINECS: 231-133-4
UN: 3208
ADR: 4.3 II
Merck Index: 12,08994

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR