VWR® INCU-Line® IL 10, IL 23, IL 56, Digitale incubatoren
Leverancier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
390-0384EA 1020 EUR
390-0384 390-0385 390-0482 390-0999
VWR® INCU-Line® IL 10, IL 23, IL 56, Digitale incubatoren
Incubatoren Compacte tafelincubatoren
IL 10, IL 23 and IL 56 digital incubators offer an economical and space saving solution for microbiology or haematology applications. The temperature can be freely adjusted in increments of 0,1 °C up to a maximum of 70 °C. Heating elements are incorporated into the housing base and walls to ensure temperature stability and uniformity. The IL 10 is available with a transparent Plexiglas® window to provide full visibility or a painted, opaque Plexiglas® window to protect photosensitive samples. IL 23 and IL 56 come as standard with a transparent Plexiglas® window. The housing and door frameMore Product Information