CO₂ incubators with hot air sterilisation and humidity regulation, CBF Series

Leverancier: Binder
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

9640-0017 9640-0018 9640-0021 9640-0022 9640-0025
390-1618EA 13020 EUR
390-1618 390-1619 390-1622 390-1623 390-1626
CO₂ incubators with hot air sterilisation and humidity regulation, CBF Series
Incubatoren CO₂-incubatoren
The CBF series CO₂ incubators are the premium series of CO₂ incubators with humidity control. Units are suitable for all sensitive incubation applications and ensure optimal cell growth in GMP environments. A CBF series CO₂ incubator can be used for complex cultivation experiments or individual environments with hypoxic conditions thanks to the comprehensive range of options and accessories. Units ensure consistent results including in cell-based assays with multiwell plates.

  • Contamination-safe humidification system without water pan
  • Tried-and-tested anti-
More Product Information
Order Now

Specificaties

Model CBF 170 CBF 260
Environmentally Preferable
Inhoud 170 L 267 L
Convectie type Mantelverwarming Mantelverwarming
Inwendige b×d×h 560×505×600 mm 620×575×750 mm
Uitwendige b×d×h 680×715×870 mm 740×785×1020 mm
Elektrisch 200/230 V 1~ 50/60 Hz 200/230 V 1~ 50/60 Hz

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR