Titanium(III,IV) oxide ≥99.9% (metals basis)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

39000.14 39000.22
39000.14EA 117 EUR
39000.14 39000.22
Titanium(III,IV) oxide ≥99.9% (metals basis)
Titanium(III,IV) oxide

Purity: 99.9% CAS No.: 12065-65-5 Molecular Formula: Ti3O5 Physical Form: 3-10mm Pieces Density: 4.20 Odour: Odourless MDL No.: MFCD00210650


Used for optical coatings

Formule: O₅Ti₃
Smeltpunt: 1800 °C
Dichtheid: 4,20
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00210650
CAS nummer: 12065-65-5

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR