Crucibles, glassy carbon
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
39011.KT 38376.KT 39003.KT 39004.KT 39005.KT 39006.KT 39008.KT 39010.KT 39012.KT 39014.KT
39011.KTEA 588 EUR
39011.KT 38376.KT 39003.KT 39004.KT 39005.KT 39006.KT 39008.KT 39010.KT 39012.KT 39014.KT
Crucibles, glassy carbon
Kroezen
These crucibles are manufactured from glassy carbon. Available in conical and cylindrical types.
  • Resistance to all wet decomposition agents
  • No contamination of analytical samples
  • Stability to acid and alkaline melts
  • No wetting effect of metal melts
  • Good resistance to thermal shock allows rapid heating and cooling times
  • Resistive to abrasive wear
  • Good electrical conductivity
Glassy carbon, a brittle form of carbon with a randomised structure, has certain specific properties making it appropriate for fields of application outside the scope of carbonMore Product Information