4-(Dimethylamino)benzaldehyde ≥99.0% (by HPLC) for the determination of hydroxyproline, Fluka™

Leverancier: Honeywell Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

39070-250G 39070-50G
39070-250GEA 238 EUR
39070-250G 39070-50G
4-(Dimethylamino)benzaldehyde ≥99.0% (by HPLC) for the determination of hydroxyproline, Fluka™
4-(Dimethylamino)benzaldehyde
Formule: (CH₃)₂NC₆H₄CHO
Kookpunt: 176…177 °C (23 hPa)
Smeltpunt: 72…75 °C
Vlampunt: 164 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003381
CAS nummer: 100-10-7
EINECS: 202-819-0
Merck Index: 13,03257
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f59709e90756a237e9c4bd808e8fd50ac9367d91&vid=909765f6b48eb3673cc290753e0ead3358b739cf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (HPLC) Min. 99.0 %
Melting point 73 - 75 °C
Loss on drying Max. 0.5 %
Colour of the solution complying
Turbidity of solution complying
Residue on ignition (as SO₄) Max. 0.1 %
Identity (NMR) complying
Application test complying
Calcium (Ca) Max. 30 ppm
Cadmium (Cd) Max. 5 ppm
Cobalt (Co) Max. 5 ppm
Chromium (Cr) Max. 5 ppm
Copper (Cu) Max. 5 ppm
Iron (Fe) Max. 10 ppm
Potassium (K) Max. 50 ppm
Magnesium (Mg) Max. 5 ppm
Manganese (Mn) Max. 5 ppm
Sodium (Na) Max. 50 ppm
Nickel (Ni) Max. 5 ppm
Lead (Pb) Max. 5 ppm
Zinc (Zn) Max. 5 ppm
Total S (as SO₄) Max. 50 ppm
Chloride (Cl) Max. 50 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR